Merck
  • Iridium-catalyzed addition of aroyl chlorides and aliphatic acid chlorides to terminal alkynes.
Iridium-catalyzed addition of aroyl chlorides and aliphatic acid chlorides to terminal alkynes.
Journal of the American Chemical Society (2011-12-14)
Tomohiro Iwai, Tetsuaki Fujihara, Jun Terao, Yasushi Tsuji
ABSTRACT
Iridium complexes show high catalytic activity in intermolecular additions of acid chlorides to terminal alkynes to afford valuable (Z)-β-chloro-α,β-unsaturated ketones. Ligands in the catalytic system play a crucial role in this reaction. An N-heterocyclic carbene (NHC) is an efficient ligand for the addition of aroyl chlorides, while dicyclohexyl(2-methylphenyl)phosphine (PCy(2)(o-Tol)) is indispensable for the reaction of aliphatic acid chlorides. The addition reactions proceed regio- and stereoselectively with suppression of decarbonylation and β-hydrogen elimination, which have been two major intrinsic problems in transition-metal-catalyzed reactions. Stoichiometric reactions of active iridium catalysts with aroyl chlorides and aliphatic acid chlorides are carried out to gain insights into the reaction mechanisms.
Sigma-Aldrich
trans-2-Phenylvinylboronic acid, 97%
Pricing and availability is not currently available.