Merck
  • ACOT12-Dependent Alteration of Acetyl-CoA Drives Hepatocellular Carcinoma Metastasis by Epigenetic Induction of Epithelial-Mesenchymal Transition.
ACOT12-Dependent Alteration of Acetyl-CoA Drives Hepatocellular Carcinoma Metastasis by Epigenetic Induction of Epithelial-Mesenchymal Transition.
Cell metabolism (2019-01-22)
Ming Lu, Wen-Wei Zhu, Xuan Wang, Jing-Jie Tang, Kai-Li Zhang, Guang-Yang Yu, Wei-Qing Shao, Zhi-Fei Lin, Sheng-Hao Wang, Lu Lu, Jian Zhou, Lian-Xin Wang, Hu-Liang Jia, Qiong-Zhu Dong, Jin-Hong Chen, Jian-Quan Lu, Lun-Xiu Qin
ABSTRACT
Metabolic reprogramming plays an important role in supporting tumor growth. However, little is known about the metabolic alterations that promote cancer metastasis. In this study, we identify acyl-CoA thioesterase 12 (ACOT12) as a key player in hepatocellular carcinoma (HCC) metastasis. The expression of ACOT12 is significantly down-regulated in HCC tissues and is closely associated with HCC metastasis and poor survival of HCC patients. Gain- and loss-of-function studies demonstrate that ACOT12 suppresses HCC metastasis both in vitro and in vivo. Further mechanistic studies reveal that ACOT12 regulates the cellular acetyl-CoA levels and histone acetylation in HCC cells and that down-regulation of ACOT12 promotes HCC metastasis by epigenetically inducing TWIST2 expression and the promotion of epithelial-mesenchymal transition. Taken together, our findings link the alteration of acetyl-CoA with HCC metastasis and imply that ACOT12 could be a prognostic marker and a potential therapeutic target for combating HCC metastasis.
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
Sigma-Aldrich
ChIPAb+ Acetyl-Histone H4 - ChIP Validated Antibody and Primer Set, serum, from rabbit
Sigma-Aldrich
Anti-acetyl-Histone H3 Antibody, from rabbit