Merck
  • Interaction between IGF2-PI3K axis and cancer-associated-fibroblasts promotes anal squamous carcinogenesis.
Interaction between IGF2-PI3K axis and cancer-associated-fibroblasts promotes anal squamous carcinogenesis.
International journal of cancer (2019-02-05)
Wulfran Cacheux, Astrid Lièvre, Sophie Richon, Sophie Vacher, Elsy El Alam, Adrien Briaux, Rania El Botty, Pascale Mariani, Bruno Buecher, Anne Schnitzler, Jorge Barbazan, Sergio Roman-Roman, Ivan Bièche, Virginie Dangles-Marie
ABSTRACT
Anal squamous cell carcinoma (ASCC) is a rare tumour, but its incidence is increasing. Standard chemoradiotherapy fails to cure 20% of patients and no targeted therapy is currently approved for recurrent ASCC. The PI3K/Akt/mTOR pathway is frequently altered in this poorly characterised carcinoma. IGF2 was identified here as a key factor in ASCC oncogenesis, as IGF2 was shown to play a crucial role in the PI3K pathway with frequent (~60%) and mutually exclusive genomic alterations in IGF2, IGF1R, PTEN and PIK3CA genes. We also demonstrated that IGF2 expression is mainly due to cancer-associated fibroblasts and that IGF2 secreted by cancer-associated fibroblasts from ASCC samples promotes proliferation of a human ASCC cell line via IGF2 paracrine signalling. Altogether, these results highlight the key role of the IGF2/PI3K axis, and the major role of cancer-associated fibroblasts in tumour growth via IGF2 secretion, suggesting a major role of IGF2/IGF1R inhibitors in ASCC therapies.