Merck
  • CDK12-mediated transcriptional regulation of noncanonical NF-κB components is essential for signaling.
CDK12-mediated transcriptional regulation of noncanonical NF-κB components is essential for signaling.
Science signaling (2018-08-02)
Kate L Henry, Debra Kellner, Bekim Bajrami, John E Anderson, Mercedes Beyna, Govinda Bhisetti, Tom Cameron, Andrew G Capacci, Andrea Bertolotti-Ciarlet, Jun Feng, Benbo Gao, Brian Hopkins, Tracy Jenkins, Kejie Li, Tricia May-Dracka, Paramasivam Murugan, Ru Wei, Weike Zeng, Norm Allaire, Alan Buckler, Christine Loh, Peter Juhasz, Brian Lucas, Katelin A Ennis, Elisabeth Vollman, Ellen Cahir-McFarland, Erik C Hett, Michelle L Ols
ABSTRACT
Members of the family of nuclear factor κB (NF-κB) transcription factors are critical for multiple cellular processes, including regulating innate and adaptive immune responses, cell proliferation, and cell survival. Canonical NF-κB complexes are retained in the cytoplasm by the inhibitory protein IκBα, whereas noncanonical NF-κB complexes are retained by p100. Although activation of canonical NF-κB signaling through the IκBα kinase complex is well studied, few regulators of the NF-κB-inducing kinase (NIK)-dependent processing of noncanonical p100 to p52 and the subsequent nuclear translocation of p52 have been identified. We discovered a role for cyclin-dependent kinase 12 (CDK12) in transcriptionally regulating the noncanonical NF-κB pathway. High-content phenotypic screening identified the compound 919278 as a specific inhibitor of the lymphotoxin β receptor (LTβR), and tumor necrosis factor (TNF) receptor superfamily member 12A (FN14)-dependent nuclear translocation of p52, but not of the TNF-α receptor-mediated nuclear translocation of p65. Chemoproteomics identified CDK12 as the target of 919278. CDK12 inhibition by 919278, the CDK inhibitor THZ1, or siRNA-mediated knockdown resulted in similar global transcriptional changes and prevented the LTβR- and FN14-dependent expression of MAP3K14 (which encodes NIK) as well as NIK accumulation by reducing phosphorylation of the carboxyl-terminal domain of RNA polymerase II. By coupling a phenotypic screen with chemoproteomics, we identified a pathway for the activation of the noncanonical NF-κB pathway that could serve as a therapeutic target in autoimmunity and cancer.
Sigma-Aldrich
Actinomycin D, from Streptomyces sp., ≥95% (HPLC)
Sigma-Aldrich
Anti-NFκB p52 Antibody, Upstate®, from mouse
Sigma-Aldrich
Amgen16, ≥98% (HPLC)