Merck
  • Quantitative Assessment of Nanoparticle Biodistribution by Fluorescence Imaging, Revisited.
Quantitative Assessment of Nanoparticle Biodistribution by Fluorescence Imaging, Revisited.
ACS nano (2018-06-20)
Fanfei Meng, Jianping Wang, Qineng Ping, Yoon Yeo
ABSTRACT
Fluorescence-based whole-body imaging is widely used in the evaluation of nanoparticles (NPs) in small animals, often combined with quantitative analysis to indicate their spatiotemporal distribution following systemic administration. An underlying assumption is that the fluorescence label represents NPs and the intensity increases with the amount of NPs and/or the labeling dyes accumulated in the region of interest. We prepare DiR-loaded poly(lactic- co-glycolic acid) (PLGA) NPs with different surface layers (polyethylene glycol with and without folate terminus) and compare the distribution of fluorescence signals in a mouse model of folate-receptor-expressing tumors by near-infrared fluorescence whole-body imaging. Unexpectedly, we observe that fluorescence distribution patterns differ far more dramatically with DiR loading than with the surface ligand, reaching opposite conclusions with the same type of NPs (tumor-specific delivery vs predominant liver accumulation). Analysis of DiR-loaded PLGA NPs reveals that fluorescence quenching, dequenching, and signal saturation, which occur with the increasing dye content and local NP concentration, are responsible for the conflicting interpretations. This study highlights the critical need for validating fluorescence labeling of NPs in the quantitative analysis of whole-body imaging. In light of our observation, we make suggestions for future whole-body fluorescence imaging in the in vivo evaluation of NP behaviors.
Sigma-Aldrich
Poly(lactide-co-glycolide)-Flamma Fluor near-IR, lactide:glycolide (50:50), Mn 20,000-30,000
Pricing and availability is not currently available.