Merck
  • Analysis of mono-phosphate nucleotides as a potential method for quantification of DNA using high performance liquid chromatography-inductively coupled plasma-mass spectrometry.
Analysis of mono-phosphate nucleotides as a potential method for quantification of DNA using high performance liquid chromatography-inductively coupled plasma-mass spectrometry.
Analytical and bioanalytical chemistry (2011-08-31)
Claire L Camp, Barry L Sharp, Helen J Reid, John Entwisle, Heidi Goenaga-Infante
ABSTRACT
The determination of total deoxyribonucleic acid (DNA) concentration is of great importance in many biological and bio-medical analyses. The quantification of DNA is traditionally performed by UV spectroscopy; however the results can be affected greatly by the sample matrix. The proposed method quantifies phosphorus in digested calf thymus DNA and human DNA by high performance liquid chromatography (HPLC) combined with inductively coupled plasma mass spectrometry (ICP-MS). The method presented showed excellent baseline separation between all four DNA mono-nucleotides and 5'UMP. The ability of LC-ICP-MS to provide an internal check that only DNA derived phosphorus was counted in the assay was demonstrated by establishing a mass balance between the total phosphorous signal from undigested DNA and that from the speciated DNA. Column recoveries ranging from 95% to 99% for phosphorus resulted in a mass balance of 95% ± 0.5% for standard nucleotides, determined by LC-ICP-MS, compared to total DNA determined by flow injection coupled to ICP-MS (FI-ICP-MS). The method for quantification was validated by analysis of NIST SRM 2,372; a total speciated DNA recovery of 52.1 ng/μL, compared with an expected value of 53.6 ng/μL, was determined by external calibration. From repeat measurements, a mass balance of 97% ± 0.5% for NIST DNA was achieved. The method limits of detection for individual nucleotides were determined between 0.8 and 1.7 μg L(-1) ((31)P) for individual nucleotides by LC-ICP-MS, and 360 ng L(-1) for 5'AMP by direct nebulisation.
Sigma-Aldrich
Deoxyribonucleic acid sodium salt from calf thymus, suitable for substrate for DNase (Readily soluble)
Pricing and availability is not currently available.
Sigma-Aldrich
Deoxyribonucleic acid sodium salt from calf thymus, Type I, fibers
Pricing and availability is not currently available.