Merck
  • Epigenetic mechanisms for silencing glutathione S-transferase m2 expression by hypermethylated specificity protein 1 binding in lung cancer.
Epigenetic mechanisms for silencing glutathione S-transferase m2 expression by hypermethylated specificity protein 1 binding in lung cancer.
Cancer (2011-01-20)
Sheau-Chung Tang, Ming-Fang Wu, Ruey-Hong Wong, Yu-Fan Liu, Lee-Chun Tang, Chien-Hung Lai, Chung-Ping Hsu, Jiunn-Liang Ko
ABSTRACT
Glutathione S-transferases M2 (GST-M2) is a detoxifying enzyme. Low expression levels of GST-M2 have been detected in lung cancer cells. However, little is known about the regulation of GST-M2 in lung cancer cells. In this study, the authors investigated the epigenetic regulatory mechanisms of GST-M2 in lung cancer cells. The authors evaluated the promoter methylation of GST-M2 in lung cancer cells after treatment with the DNA methyltransferase (DNMT) inhibitor 5'-aza-2'-deoxycytidine (5'-aza-dC). Reporter activity assays, chromatin immunoprecipitation (ChIP), electrophoretic mobility-shift assays, and small interfering RNA (siRNA) assays were used to determine whether the methylation of specificity protein 1 (Sp1) affected binding to the GST-M2 promoter or regulated GST-M2 transcription. Real-time polymerase chain reaction was used to determine GST-M2 and DNMT-3b messenger RNA levels in 73 nonsmall cell lung cancer (NSCLC) tissues. GST-M2 expression was restored after treatment with 5'-aza-dC in lung cancer cells. GST-M2 exhibited high frequency of promoter hypermethylation in lung cancer cells and NSCLC tumor tissues. CpG hypermethylation abated Sp1 binding to the GST-M2 promoter in lung cancer. Knockdown of Sp1 in normal lung cells reduced GST-M2 expression, and silencing of DNMT-3b increased GST-M2 expression in lung cancer cells. In addition, DNMT-3b expression was significantly higher in lung tumors with low levels of GST-M2 expression than in lung tumors with high levels of GST-M2 expression, especially among women and among patients who had stage I disease. Epigenetic silencing of GST-M2 was distinguished from Sp1-mediated GST-M2 transcriptional expression. The authors concluded that this represents a mechanism that leads to decreased expression of GST-M2 in lung cancer cells.
Sigma-Aldrich
5-Aza-2′-deoxycytidine, ≥97%
Pricing and availability is not currently available.