Merck
  • Label-free cytosensing of cancer cells based on the interaction between protein and an electron-transfer carbohydrate-mimetic peptide.
Label-free cytosensing of cancer cells based on the interaction between protein and an electron-transfer carbohydrate-mimetic peptide.
Analytica chimica acta (2018-10-18)
Kazuharu Sugawara, Hideki Kuramitz, Toshihiko Kadoya
ABSTRACT
We used an electron-transfer carbohydrate-mimetic peptide (YYYYC) to construct an electrochemical cytosensing system. Magnetic beads were modified with either asialofetuin (ASF) or soybean agglutinin (SBA) to evaluate the effect on cell sensing. Because SBA binds to the galactose residue that exists at the terminals of the carbohydrate chains in ASF, the target protein was accumulated on the protein magnetic beads. SBA is an example of N-acetylgalactosamine- and galactose-binding proteins that readily combine with YYYYC. When the peptides and protein-immobilized beads competed for a target protein, the peak current of the peptides changed according to the concentration of the protein at the 10-12 M level. Next, human myeloid leukemia cells (K562 cell) were measured using the peptide and the carbohydrate chains on the cell surface that recognize SBA. The electrode response was linear to the number of K562 cells and ranged from 1.0 × 102 to 5.0 × 103 cells mL-1. In addition, detection of a human liver cancer cell (HepG2 cell) was carried out using interactions with the peptide, the ASF receptors in HepG2 cells, and the carbohydrate chains of ASF. The peak currents were proportional and ranged between 5.0 × 101 and 1.5 × 103 cells mL-1. When the values estimated from an electrochemical process were compared with those obtained by ELISA, the results were within the acceptable range of measurement error.