Merck
(Rp)-8-pCPT-cGMPS, a novel cGMP-dependent protein kinase inhibitor.
European journal of pharmacology (1994-10-14)
E Butt, M Eigenthaler, H G Genieser
ABSTRACT
In the present study, the inhibitory effect of the cGMP analog (Rp)-8-(para-chlorophenylthio)guanosine-3',5'-cyclic monophosphorothioate ((Rp)-8-pCPT-cGMPS) on the cGMP-dependent protein kinase-mediated protein phosphorylation in intact human platelets was investigated. In vitro phosphorylation experiments with the substrate kemptide demonstrated an inhibition of the cGMP-dependent protein kinase by (Rp)-8-pCPT-cGMPS with a Ki of 0.5 microM. In intact human platelets, (Rp)-8-pCPT-cGMPS antagonized the activation of the cGMP-dependent protein kinase by 8-pCPT-cGMP without affecting cAMP-dependent protein kinase or cGMP-regulated phosphodiesterases. The data obtained suggest that (Rp)-8-pCPT-cGMPS may be a useful tool for studying the role of cGMP in vitro and in intact cells.
Sigma-Aldrich
8-(4-Chlorophenylthio)-guanosine 3′,5′-cyclic monophosphorothioate, Rp Isomer triethylammonium salt, ≥98% (HPLC), solid