Merck
  • Tuned Density of Anti-Tissue Factor Antibody Fragment onto siRNA-Loaded Polyion Complex Micelles for Optimizing Targetability into Pancreatic Cancer Cells.
Tuned Density of Anti-Tissue Factor Antibody Fragment onto siRNA-Loaded Polyion Complex Micelles for Optimizing Targetability into Pancreatic Cancer Cells.
Biomacromolecules (2018-05-17)
Hyun Su Min, Hyun Jin Kim, Jooyeon Ahn, Mitsuru Naito, Kotaro Hayashi, Kazuko Toh, Beob Soo Kim, Yasuhiro Matsumura, Ick Chan Kwon, Kanjiro Miyata, Kazunori Kataoka
ABSTRACT
Antibody fragment (Fab')-installed polyion complex (PIC) micelles were constructed to improve targetability of small interfering RNA (siRNA) delivery to pancreatic cancer cells. To this end, we synthesized a block copolymer of azide-functionalized poly(ethylene glycol) and poly(l-lysine) and prepared PIC micelles with siRNA. Then, a dibenzylcyclooctyne (DBCO)-modified antihuman tissue factor (TF) Fab' was conjugated to azido groups on the micellar surface. A fluorescence correlation spectroscopic analysis revealed that 1, 2, or 3 molecule(s) of Fab'(s) were installed onto one micellar nanoparticle according to the feeding ratio of Fab' (or DBCO) to micelle (or azide). The resulting micelles exhibited ∼40 nm in hydrodynamic diameter, similar to that of the parent micelles before Fab' conjugation. Flow cytometric analysis showed that three molecules of Fab'-installed PIC micelles (3(Fab')-micelles) had the highest binding affinity to cultured pancreatic cancer BxPC3 cells, which are known to overexpress TF on their surface. The 3(Fab')-micelles also exhibited the most efficient gene silencing activity against polo-like kinase 1 mRNA in the cultured cancer cells. Furthermore, the 3(Fab')-micelles exhibited high penetrability and the highest cellular internalization amounts in BxPC3 spheroids compared with one or two molecule(s) of Fab'-installed PIC micelles. These results demonstrate the potential of anti-TF Fab'-installed PIC micelles for active targeting of stroma-rich pancreatic tumors.
Sigma-Aldrich
Dimethyl 3,3′-dithiopropionimidate dihydrochloride, powder
Sigma-Aldrich
2-Iminothiolane hydrochloride, ≥98% (TLC), powder