- Structural mimicry of the α-helix in aqueous solution with an isoatomic α/β/γ-peptide backbone.
Structural mimicry of the α-helix in aqueous solution with an isoatomic α/β/γ-peptide backbone.
Journal of the American Chemical Society (2011-04-28)
Tomohisa Sawada, Samuel H Gellman
PMID21520956
ABSTRACT
Artificial mimicry of α-helices offers a basis for development of protein-protein interaction antagonists. Here we report a new type of unnatural peptidic backbone, containing α-, β-, and γ-amino acid residues in an αγααβα repeat pattern, for this purpose. This unnatural hexad has the same number of backbone atoms as a heptad of α residues. Two-dimensional NMR data clearly establish the formation of an α-helix-like conformation in aqueous solution. The helix formed by our 12-mer α/β/γ-peptide is considerably more stable than the α-helix formed by an analogous 14-mer α-peptide, presumably because of the preorganized β and γ residues employed.
Sigma-Aldrich
O-(2-Oxo-1(2H)pyridyl)-N,N,N′,N′-tetramethyluronium tetrafluoroborate, ≥99.0% (HPLC)