Merck
  • The Rab11-binding protein RELCH/KIAA1468 controls intracellular cholesterol distribution.
The Rab11-binding protein RELCH/KIAA1468 controls intracellular cholesterol distribution.
The Journal of cell biology (2018-03-09)
Tomoaki Sobajima, Shin-Ichiro Yoshimura, Tomomi Maeda, Haruhiko Miyata, Eiji Miyoshi, Akihiro Harada
ABSTRACT
Cholesterol, which is endocytosed to the late endosome (LE)/lysosome, is delivered to other organelles through vesicular and nonvesicular transport mechanisms. In this study, we discuss a novel mechanism of cholesterol transport from recycling endosomes (REs) to the trans-Golgi network (TGN) through RELCH/KIAA1468, which is newly identified in this study as a Rab11-GTP- and OSBP-binding protein. After treating cells with 25-hydroxycholesterol to induce OSBP relocation from the cytoplasm to the TGN, REs accumulated around the TGN area, but this accumulation was diminished in RELCH- or OSBP-depleted cells. Cholesterol content in the TGN was decreased in Rab11-, RELCH-, and OSBP-depleted cells and increased in the LE/lysosome. According to in vitro reconstitution experiments, RELCH tethers Rab11-bound RE-like and OSBP-bound TGN-like liposomes and promotes OSBP-dependent cholesterol transfer from RE-like to TGN-like liposomes. These data suggest that RELCH promotes nonvesicular cholesterol transport from REs to the TGN through membrane tethering.
Sigma-Aldrich
Allyl methyl sulfone, 96%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, pH 7, ≥98%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Cycloheximide, Biotechnology Performance Certified
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
MISSION® esiRNA, targeting human OSBP