Merck
  • Isolation and partial characterization of an elastase-type enzyme from human arterial wall by lima-bean trypsin inhibitor affinity chromatography.
Isolation and partial characterization of an elastase-type enzyme from human arterial wall by lima-bean trypsin inhibitor affinity chromatography.
Artery (1980-01-01)
G Bellon, T Ooyama, W Hornebeck, L Robert
ABSTRACT
A serine protease active on insoluble elastin at neutral pH has been isolated from human aortic media employing a Lima-bean trypsin inhibitor - Sepharose column. It is also hydrolyzed Suc (Ala)3 pna and casein but was found inactive against Benzoyl-Tyr-pna and Benzoyl-Arg-pna. Its apparent molecular weight as determined by SDS-PAGE was 22,300 Daltons. It differs from other elastases of human origin (human pancreatic elastase-1, human pancreatic elastase-2, human leucocyte elastase) on the basis of amino-acid composition and immunological specificity.
Sigma-Aldrich
Trypsin inhibitor from Phaseolus limensis (lima bean), Type II-L, crude powder
Pricing and availability is not currently available.