Merck
  • Regenerative potential of human adipose-derived stromal cells of various origins.
Regenerative potential of human adipose-derived stromal cells of various origins.
Journal of cranio-maxillo-facial surgery : official publication of the European Association for Cranio-Maxillo-Facial Surgery (2015-11-07)
Susanne Jung, Benedikt Kleineidam, Johannes Kleinheinz
ABSTRACT
In regenerative concepts, the potential of adult stem cells holds great promise concerning an individualized therapeutic approach. These cells provide renewable progenitor cells to replace aged tissue, and play a significant role in tissue repair and regeneration. In this investigation, the characteristics of different types of adipose tissue are analysed systematically with special attention to their proliferation and differentiation potential concerning the angiogenic and osteogenic lineage. Tissue samples from subcutaneous, visceral, and omental fat were processed according to standard procedures. The cells were characterized and cultivated under suitable conditions for osteogenic and angiogenic cell culture. The development of the different cell cultures as well as their differentiation were analysed morphologically and immunohistochemically from cell passages P1 to P12. Harvesting and isolation of multipotent cells from all three tissue types could be performed reproducibly. The cultivation of these cells under osteogenic conditions led to a morphological and immunohistochemical differentiation; mineralization could be detected. The most stable results were observed for the cells of subcutaneous origin. An osteogenic differentiation from adipose-derived cells from all analysed fatty tissues can be achieved easily and reproducibly. In therapeutic concepts including angiogenic regeneration, adipose-derived cells from subcutaneous tissue provide the optimal cellular base.
Sigma-Aldrich
Anti-von Willebrand Factor antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution