![TSKgel® Super-Phenyl HPLC Column phase phenyl, L × I.D. 5 cm × 2 mm, 2.3 μm particle size](/deepweb/assets/sigmaaldrich/product/images/140/085/e82012d0-d8eb-40f7-a6b6-51875ab5def7/640/e82012d0-d8eb-40f7-a6b6-51875ab5def7.jpg)
TSKgel® Super-Phenyl (2.3 µm) HPLC Columns
![TSKgel® Super-Phenyl HPLC Column phase phenyl, L × I.D. 5 cm × 2 mm, 2.3 μm particle size](/deepweb/assets/sigmaaldrich/product/images/140/085/e82012d0-d8eb-40f7-a6b6-51875ab5def7/640/e82012d0-d8eb-40f7-a6b6-51875ab5def7.jpg)
Synonym(s):
TSKgel® Reversed Phase HPLC Column
Products
Product Category
Manufacturer
Product Line
Agency
USP Suitability
Separation Technique
ID (mm)
Length (cm)
Particle Size (µm)
Pore Size (Å)
Technique
Showing 1-5 of 5
Compare
Product Number
Description
Pricing