Skip to Content
Merck
TSKgel® Q-STAT HPLC Column phase ammonium ion, quaternary, L × I.D. 3.5 cm × 3 mm, 10 μm particle size

TSKgel® Q-STAT (10 µm) HPLC Column

TSKgel® Q-STAT HPLC Column phase ammonium ion, quaternary, L × I.D. 3.5 cm × 3 mm, 10 μm particle size
Synonym(s):
TSKgel® Q-STAT Columns

Products

Product Category
Manufacturer
Product Line
Agency
USP Suitability
Separation Technique
ID (mm)
Length (cm)
Particle Size (µm)
Technique
Showing 1-1 of 1