Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)manganese(III)
Synonym(s):
Mn(TMHD)3, Tris(dipivaloylmethanato)manganese
Linear Formula:
Mn(C11H19O2)3
CAS Number:
Molecular Weight:
604.74
Products
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Reagent Type
Core
Showing 1-1 of 1
Compare
Product Number
Description
Pricing