Skip to Content
Merck
Triamterene Related Compound C Pharmaceutical Secondary Standard; Certified Reference Material

Triamterene Related Compound C

Triamterene Related Compound C Pharmaceutical Secondary Standard; Certified Reference Material
Synonym(s):
2,4-Diamino-7-hydroxy-6-phenylpteridine, NSC 33416
Empirical Formula (Hill Notation):
C12H10N6O
CAS Number:
Molecular Weight:
254.25

Products

Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Technique