Skip to Content
Merck
trans-2-Hexen-1-ol 96%

trans-2-Hexen-1-ol

trans-2-Hexen-1-ol 96%
Linear Formula:
CH3CH2CH2CH=CHCH2OH
CAS Number:
Molecular Weight:
100.16
EC Number:

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Agency
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing