Topiramate-D12 solution
Empirical Formula (Hill Notation):
C12D12H9NO8S
CAS Number:
Molecular Weight:
351.44
Products
Brand
Formula Weight
Manufacturer
Physical Form
Format
Quality Segment
Application
Package Size
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2