
Tioconazole Related Compound A

Synonym(s):
1-[2-(2,4-Dichlorophenyl)-2-(3-thienylmethoxy)ethyl]-1H-imidazole monohydrochloride, 1-{2,4-Dichloro-β-[(3-thenyl)oxy]phenethyl}imidazole hydrochloride
Empirical Formula (Hill Notation):
C16H14Cl2N2OS · HCl
CAS Number:
Molecular Weight:
389.73
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing