tert-Butyldimethylsilyl chloride solution
Synonym(s):
tert-Butylchlorodimethylsilane solution, tert-Butyldimethylchlorosilane, tert-Butyldimethylsilyl chloride, TBDMSCl solution
Linear Formula:
(CH3)3CSi(CH3)2Cl
CAS Number:
Molecular Weight:
150.72
Products
Brand
Formula Weight
Physical Form
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing