Skip to Content
IL
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Specially Denatured Alcohol
Synonym(s):
Ethanol, SDA
CAS Number:
64-17-5
Products
Product Category
solvents (18)
Brand
Sigma-Aldrich (18)
Physical Form
Application
Filter Products
Showing 1-18 of 18
Compare
Product Number
Description
Pricing
792985
Specially Denatured Alcohol,
200 proof, SDA 3A, contains Methanol
Expand
View Pricing
793043
Specially Denatured Alcohol,
190 proof, SDA 23A, contains Acetone
Expand
View Pricing
792896
Specially Denatured Alcohol,
200 proof, SDA 2B-3, contains Toluene
Expand
View Pricing
793116
Specially Denatured Alcohol,
200 proof, SDA 39C, contains Diethyl phthalate
Expand
View Pricing
793132
Specially Denatured Alcohol,
200 proof, SDA 40 (40-2), contains 0.14 % (v/v)
tert
-Butyl alcohol and Brucine sulfate
Expand
View Pricing
793108
Specially Denatured Alcohol,
190 proof, SDA 35A, contains Ethyl acetate
Expand
View Pricing
793140
Specially Denatured Alcohol,
190 proof, SDA 40 (40-2), contains 0.14 % (v/v)
tert
-Butyl alcohol and Brucine sulfate
Expand
View Pricing
793124
Specially Denatured Alcohol,
190 proof, SDA 39C, contains Diethyl phthalate
Expand
View Pricing
792918
Specially Denatured Alcohol,
190 proof, SDA 2B-3, contains Toluene
Expand
View Pricing
793078
Specially Denatured Alcohol,
190 proof, SDA 30, contains Methanol
Expand
View Pricing
793051
Specially Denatured Alcohol,
200 proof, SDA 30, contains Methanol
Expand
View Pricing
793094
Specially Denatured Alcohol,
200 proof, SDA 35A, contains Ethyl acetate
Expand
View Pricing
792969
Specially Denatured Alcohol,
200 proof, SDA 2B-5, contains n-Hexane
Expand
View Pricing
792926
Specially Denatured Alcohol,
200 proof, SDA 2B-4, contains Heptanes
Expand
View Pricing
792942
Specially Denatured Alcohol,
200 proof, SDA 2B-4, contains n-Heptane
Expand
View Pricing
792950
Specially Denatured Alcohol,
190 proof, SDA 2B-4, contains n-Heptane
Expand
View Pricing
792977
Specially Denatured Alcohol,
190 proof, SDA 2B-5, contains n-Hexane
Expand
View Pricing
792934
Specially Denatured Alcohol,
190 proof, SDA 2B-4, contains Heptanes
Expand
View Pricing