![Solvent Yellow 124 analytical standard](/deepweb/assets/sigmaaldrich/product/structures/271/701/cadaac64-9cb1-4797-8471-55f7b06c92fb/640/cadaac64-9cb1-4797-8471-55f7b06c92fb.png)
Solvent Yellow 124
![Solvent Yellow 124 analytical standard](/deepweb/assets/sigmaaldrich/product/structures/271/701/cadaac64-9cb1-4797-8471-55f7b06c92fb/640/cadaac64-9cb1-4797-8471-55f7b06c92fb.png)
Synonym(s):
N-Ethyl-N-[2-(1-isobutoxyethoxy)ethyl]-p-(phenylazo)aniline
Empirical Formula (Hill Notation):
C22H31N3O2
Products
Brand
Formula Weight
Purity
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing