Skip to Content
Merck
Sodium dodecyl-d25 sulfate ≥98 atom % D, ≥98% (CP)

Sodium dodecyl-d25 sulfate

Sodium dodecyl-d25 sulfate ≥98 atom % D, ≥98% (CP)
Synonym(s):
Dodecyl-d25 sulfate sodium salt, SDS-d25, sodium lauryl sulfate-d25
Linear Formula:
CD3(CD2)11OSO3Na
CAS Number:
Molecular Weight:
313.53

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Purity
Isotope
Markush Class
Markush Group
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing