Skip to Content
Merck
Protopine phyproof® Reference Substance

Protopine

Protopine phyproof® Reference Substance
Synonym(s):
4,6,7,14-Tetrahydro-5-methylbis[1,3]benzodioxolo[4,5-c:5′,6′-g]azecin-13(5H)-one
Empirical Formula (Hill Notation):
C20H19NO5
CAS Number:
Molecular Weight:
353.37

Products

Formula Weight
Manufacturer
Product Line
Format
Application
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing