![Primaquine Phosphate pharmaceutical secondary standard, certified reference material](/deepweb/assets/sigmaaldrich/product/structures/231/045/9337b14d-a16f-4d54-9910-3f5606609df0/640/9337b14d-a16f-4d54-9910-3f5606609df0.png)
Primaquine phosphate
![Primaquine Phosphate pharmaceutical secondary standard, certified reference material](/deepweb/assets/sigmaaldrich/product/structures/231/045/9337b14d-a16f-4d54-9910-3f5606609df0/640/9337b14d-a16f-4d54-9910-3f5606609df0.png)
Synonym(s):
Primaquine bisphosphate, 8-(4-Amino-1-methylbutylamino)-6-methoxyquinoline diphosphate salt, Primaquine diphosphate salt
Empirical Formula (Hill Notation):
C15H21N3O · 2H3PO4
CAS Number:
Molecular Weight:
455.34
Products
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Package Size
API Family
Special Grade
Showing 1-2 of 2