Skip to Content
Merck
vibrant-m

Pravastatin 1,1,3,3-tetramethylbutylamine for assay

vibrant-m
Synonym(s):
Pravastatin 1,1,3,3-tetramethylbutylamine
Empirical Formula (Hill Notation):
C23H36O7 · C8H19N
CAS Number:
Molecular Weight:
553.77

Products

Formula Weight
Manufacturer
API Family
Reference Material Type
Showing 1-1 of 1