![Plate-Count-Agar (Agar 15 g/L) suitable for microbiology, NutriSelect® Plus](/deepweb/assets/sigmaaldrich/product/images/281/299/8d3423c0-e067-4907-82a9-e042925c00d0/640/8d3423c0-e067-4907-82a9-e042925c00d0.jpg)
Plate-Count-Agar (Agar 15 g/L)
![Plate-Count-Agar (Agar 15 g/L) suitable for microbiology, NutriSelect® Plus](/deepweb/assets/sigmaaldrich/product/images/281/299/8d3423c0-e067-4907-82a9-e042925c00d0/640/8d3423c0-e067-4907-82a9-e042925c00d0.jpg)
Synonym(s):
Plate-Count-Agar according to Buchbinder et al., Casein-peptone Dextrose Yeast Agar according to Buchbinder et al., Standard Methods Agar according to Buchbinder et al., Tryptone Glucose Yeast Extract Agar according to Buchbinder et al.
Products
Product Category
Brand
pH Value
Physical Form
Agency
Chemical Composition
Quality Segment
Application
Microorganism Suitability
Sterility
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing