Skip to Content
Merck
vibrant-m

Plasmid DNA Fragments of MON 810 Maize

vibrant-m
Synonym(s):
Maize GMO Standard ERM-BF413, GMO Standard ERM-BF413, Maize MON 810

Products

Manufacturer
Agency
Format
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing