Pioglitazone hydrochloride
Synonym(s):
5-[[4-[2-(5-Ethyl-2-pyridinyl)ethoxy]phenyl]methyl]-2,4-thiazolidinedione monohydrochloride, Pioglitazone hydrochloride, 5-[4-[2-(5-Ethyl-2-pyridinyl)ethoxy]benzyl]thiazolidine-2,4-dione hydrochloride
Empirical Formula (Hill Notation):
C19H20N2O3S · HCl
CAS Number:
Molecular Weight:
392.90
Products
Product Category
Brand
Color
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Physical Form
Purity
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Technique
Showing 1-5 of 5
Compare
Product Number
Description
Pricing