![Phosphodiesterase I from Crotalus atrox (Western Diamondback Rattlesnake) Type IV, crude dried venom](/deepweb/assets/sigmaaldrich/product/images/417/501/59396621-710b-49a9-80a3-efa473b6b0a8/640/59396621-710b-49a9-80a3-efa473b6b0a8.jpg)
Phosphodiesterase I from Crotalus atrox (Western Diamondback Rattlesnake)
![Phosphodiesterase I from Crotalus atrox (Western Diamondback Rattlesnake) Type IV, crude dried venom](/deepweb/assets/sigmaaldrich/product/images/417/501/59396621-710b-49a9-80a3-efa473b6b0a8/640/59396621-710b-49a9-80a3-efa473b6b0a8.jpg)
Synonym(s):
5′-Exonuclease, Oligonucleate 5′-nucleotidohydrolase
EC Number:
Products
Brand
Physical Form
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing