Phentolamine mesilate for system suitability
Synonym(s):
Phentolamine methanesulfonate salt, 2-(N-[m-Hydroxyphenyl]-p-toluidinomethyl)imidazoline
Linear Formula:
C17H19N3O · CH3SO3H
CAS Number:
Molecular Weight:
377.46
Products
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing