![O-Phospho-L-tyrosine certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland](/deepweb/assets/sigmaaldrich/product/structures/140/674/a259683f-bfde-4dd1-b7ff-ac14f48fcc51/640/a259683f-bfde-4dd1-b7ff-ac14f48fcc51.png)
O-Phospho-L-tyrosine
![O-Phospho-L-tyrosine certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland](/deepweb/assets/sigmaaldrich/product/structures/140/674/a259683f-bfde-4dd1-b7ff-ac14f48fcc51/640/a259683f-bfde-4dd1-b7ff-ac14f48fcc51.png)
Synonym(s):
L-3-(4-Hydroxyphenyl)alanine 4′-phosphate, L-Tyrosine-O-phosphate
Empirical Formula (Hill Notation):
C9H12NO6P
CAS Number:
Molecular Weight:
261.17
Products
Product Category
Brand
Color
Formula Weight
Manufacturer
Physical Form
Product Line
Quality Segment
Application
Optical Isomer
Amino Acid
Special Grade
Showing 1-2 of 2