Neomycin sulphate
Empirical Formula (Hill Notation):
C23H46N6O13 · xH2SO4
CAS Number:
Molecular Weight:
614.64 (free base basis)
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing