![N-Boc-1,3-propanediamine ≥97.0% (GC/NT)](/deepweb/assets/sigmaaldrich/product/structures/266/989/bf21b2ca-a28f-4d98-89f9-eb58ad079d37/640/bf21b2ca-a28f-4d98-89f9-eb58ad079d37.png)
N-Boc-1,3-propanediamine
![N-Boc-1,3-propanediamine ≥97.0% (GC/NT)](/deepweb/assets/sigmaaldrich/product/structures/266/989/bf21b2ca-a28f-4d98-89f9-eb58ad079d37/640/bf21b2ca-a28f-4d98-89f9-eb58ad079d37.png)
Synonym(s):
N-Boc-1,3-diaminopropane, tert-Butyl N-(3-aminopropyl)carbamate
Linear Formula:
(CH3)3COCONH(CH2)3NH2
CAS Number:
Molecular Weight:
174.24
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Reagent Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing