![mPEG40K-Succinimidyl Carboxymethyl Ester average Mn 40,000](/deepweb/assets/sigmaaldrich/product/structures/103/179/d45789d1-849d-40a1-bed7-09488b737ca4/640/d45789d1-849d-40a1-bed7-09488b737ca4.png)
mPEG40K
![mPEG40K-Succinimidyl Carboxymethyl Ester average Mn 40,000](/deepweb/assets/sigmaaldrich/product/structures/103/179/d45789d1-849d-40a1-bed7-09488b737ca4/640/d45789d1-849d-40a1-bed7-09488b737ca4.png)
Synonym(s):
mPEG-MAL, mPEG-Succinimidyl Carboxymethyl Ester
Linear Formula:
(CH3O(CH2CH2O)nCH2COOC4H4O2N
Products
Brand
Physical Form
Functionality
Shape
Polymer Composition
Polymer Type
Reactivity
Reagent Type
Showing 1-2 of 2
Compare
Product Number
Description
Pricing