Skip to Content
Merck
Monoclonal Anti-Rabbit Immunoglobulins–Biotin antibody produced in mouse clone RG-16, purified immunoglobulin, buffered aqueous solution

Monoclonal Anti-Rabbit Immunoglobulins–Biotin antibody produced in mouse

Monoclonal Anti-Rabbit Immunoglobulins–Biotin antibody produced in mouse clone RG-16, purified immunoglobulin, buffered aqueous solution
Synonym(s):
Monoclonal Anti-Rabbit Immunoglobulins (IgG, IgA, IgM)

Products

Product Category
Brand
Biological Source
Clonality
Isotype
Physical Form
Antibody Form
Research Area
Conjugate
Post Translational Modification
Non-reactive Species
Species Reactivity
Technique