Skip to Content
Merck
Monoclonal Anti-PGR antibody produced in mouse clone 4E9, purified immunoglobulin, buffered aqueous solution

Monoclonal Anti-PGR antibody produced in mouse

Monoclonal Anti-PGR antibody produced in mouse clone 4E9, purified immunoglobulin, buffered aqueous solution
Synonym(s):
Anti-NR3C3, Anti-PR

Products

Product Category
Brand
Biological Source
Clonality
Gene Alias
Isotype
Physical Form
Antibody Form
Research Area
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique