
Monoclonal Anti-I-CAM-1 (CD54) antibody produced in mouse

Synonym(s):
Monoclonal Anti-CD54, Anti-I-CAM-1
Products
Product Category
Brand
Biological Source
Clonality
Gene Alias
Isotype
Antibody Form
Research Area
Conjugate
Post Translational Modification
Species Reactivity
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing