![Monoclonal ANTI-FLAG® M2 antibody produced in mouse clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)](/deepweb/assets/sigmaaldrich/product/images/352/356/133c584d-3f4f-4aab-a3c9-b357c2480c20/640/133c584d-3f4f-4aab-a3c9-b357c2480c20.jpg)
Monoclonal ANTI-FLAG® M2 antibody produced in mouse
![Monoclonal ANTI-FLAG® M2 antibody produced in mouse clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)](/deepweb/assets/sigmaaldrich/product/images/352/356/133c584d-3f4f-4aab-a3c9-b357c2480c20/640/133c584d-3f4f-4aab-a3c9-b357c2480c20.jpg)
Synonym(s):
Anti-ddddk, Anti-dykddddk
Products
Product Category
Brand
Biological Source
Clonality
Isotype
Physical Form
Antibody Form
Conjugate
Quality Segment
Species Reactivity
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing