
Monoclonal Anti-CD16−FITC antibody produced in mouse

Synonym(s):
Anti-FcgammaRIII, Anti-FCGR3A, Anti-FcγRIII
Products
Product Category
Brand
Biological Source
Clonality
Gene Alias
Isotype
Physical Form
Antibody Form
Research Area
Conjugate
Post Translational Modification
Species Reactivity
Technique
Showing 1-4 of 4
Compare
Product Number
Description
Pricing