![Methyl cyclopropanecarboxylate for synthesis](/deepweb/assets/sigmaaldrich/product/images/101/019/e4e06131-84c9-46e2-b0e2-6fa8a3d3ca2e/640/e4e06131-84c9-46e2-b0e2-6fa8a3d3ca2e.jpg)
Methyl cyclopropanecarboxylate
![Methyl cyclopropanecarboxylate for synthesis](/deepweb/assets/sigmaaldrich/product/images/101/019/e4e06131-84c9-46e2-b0e2-6fa8a3d3ca2e/640/e4e06131-84c9-46e2-b0e2-6fa8a3d3ca2e.jpg)
Synonym(s):
Methyl cyclopropanecarboxylate
Empirical Formula (Hill Notation):
C5H8O2
CAS Number:
Molecular Weight:
100.12
Products
Brand
Formula Weight
Physical Form
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing