Meclofenamate Sodium
Synonym(s):
Meclofenamic acid sodium salt, 2-[(2,6-Dichloro-3-methylphenyl)amino]benzoic acid sodium salt
Linear Formula:
C14H10Cl2NO2Na
CAS Number:
Molecular Weight:
318.13
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2