![L-Sulforaphane ≥95% (HPLC), oil](/deepweb/assets/sigmaaldrich/product/structures/378/513/0201a22f-d956-4227-a892-dfc4ca5b181e/640/0201a22f-d956-4227-a892-dfc4ca5b181e.png)
L-Sulforaphane
![L-Sulforaphane ≥95% (HPLC), oil](/deepweb/assets/sigmaaldrich/product/structures/378/513/0201a22f-d956-4227-a892-dfc4ca5b181e/640/0201a22f-d956-4227-a892-dfc4ca5b181e.png)
Synonym(s):
(R)-1-Isothiocyanato-4-(methylsulfinyl)butane, 4-Methylsulfinylbutyl isothiocyanate
Empirical Formula (Hill Notation):
C6H11NOS2
CAS Number:
Molecular Weight:
177.29
Products
Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Quality Segment
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing