Skip to Content
Merck
Lipopolysaccharides (rough strains) from Salmonella enterica serotype minnesota Re 595 (Re mutant)

Lipopolysaccharides (rough strains) from Salmonella enterica serotype minnesota Re 595 (Re mutant)

Lipopolysaccharides (rough strains) from Salmonella enterica serotype minnesota Re 595 (Re mutant)
Synonym(s):
LPS

Products

Product Category
Brand
Biological Source
Color
Physical Form
Quality Segment
Showing 1-1 of 1