![Keracyanin chloride analytical standard](/deepweb/assets/sigmaaldrich/product/structures/284/120/6383645f-eabe-4868-be4c-38da7b292dc3/640/6383645f-eabe-4868-be4c-38da7b292dc3.png)
Keracyanin chloride
![Keracyanin chloride analytical standard](/deepweb/assets/sigmaaldrich/product/structures/284/120/6383645f-eabe-4868-be4c-38da7b292dc3/640/6383645f-eabe-4868-be4c-38da7b292dc3.png)
Synonym(s):
Cyanidin-3-O-rutinoside chloride
Empirical Formula (Hill Notation):
C27H31ClO15
Products
Brand
Formula Weight
Purity
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing