![Isorhamnetin 3-rutinoside phyproof® Reference Substance](/deepweb/assets/sigmaaldrich/product/structures/259/386/a3a50eef-ab82-4cc1-a9f5-68ff953ea53a/640/a3a50eef-ab82-4cc1-a9f5-68ff953ea53a.png)
Isorhamnetin 3-rutinoside
![Isorhamnetin 3-rutinoside phyproof® Reference Substance](/deepweb/assets/sigmaaldrich/product/structures/259/386/a3a50eef-ab82-4cc1-a9f5-68ff953ea53a/640/a3a50eef-ab82-4cc1-a9f5-68ff953ea53a.png)
Synonym(s):
Narcissoside, 3,4′,5,7-Tetrahydroxy 3′-methoxyflavone 3-rutinoside, 3-O-Methylquercetin 3-rutinoside, 3-Rutinosylisorhamnetin, Isorhamnetin 3-O-rutinoside, Isorhamnetin 3-rhamnoglucoside, Isorhamnetol 3-rutinoside, Narcissin, Quercetin 3′-methyl ether 3-rutinoside
Empirical Formula (Hill Notation):
C28H32O16
CAS Number:
Molecular Weight:
624.54
Products
Formula Weight
Manufacturer
Product Line
Purity
Format
Application
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing