
Isoforskolin

Synonym(s):
6-Acetyl-7-deacetylforskolin, 6β-Acetoxy-1α,7β,9α-trihydroxy-8,13-epoxy-labd-14-en-11-one, Isoforskolin
Empirical Formula (Hill Notation):
C22H34O7
CAS Number:
Molecular Weight:
410.50
Products
Brand
Color
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Quality Segment
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing