Skip to Content
Merck
Iron disilicide powder, -20 mesh, 99.9% trace metals basis

Iron disilicide

Iron disilicide powder, -20 mesh, 99.9% trace metals basis
Synonym(s):
Iron silicide
Empirical Formula (Hill Notation):
FeSi2
CAS Number:
Molecular Weight:
112.02

Products

Brand
Sustainability
Feature
Formula Weight
Melting Point (°C)
Physical Form
Purity
Showing 1-1 of 1
Compare
Product Number
Description
Pricing